missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Methylthiomethyl)phosphonate 97.0+%, TCI America™

Catalog No. p-7132601
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12085G 5 g
M120825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12085G Supplier TCI America Supplier No. M12085G

Chemical Identifiers

CAS 28460-01-7
Molecular Formula C6H15O3PS
Molecular Weight (g/mol) 198.217
MDL Number MFCD00015131
InChI Key RAABCCZMKIIGJW-UHFFFAOYSA-N
Synonym diethyl methylthiomethylphosphonate, diethyl methylthiomethyl phosphonate, diethyl methylthio methyl phosphonate, diethyl methylsulfanyl methylphosphonate, methylthiomethyl phosphonic acid diethyl ester, 1-ethoxy methylsulfanylmethyl phosphoryl oxyethane, phosphonic acid, methylthio methyl-, diethyl ester, diethoxy methylthiomethyl phosphino-1-one, pubchem12686
PubChem CID 520377
IUPAC Name 1-[ethoxy(methylsulfanylmethyl)phosphoryl]oxyethane
SMILES CCOP(=O)(CSC)OCC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Methylthiomethyl)phosphonate