missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Methyloxalacetate 96.0+%, TCI America™

Catalog No. M038125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M038125G Supplier TCI America Supplier No. M038125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 759-65-9
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.206
MDL Number MFCD00009163
InChI Key OQOCQBJWOCRPQY-UHFFFAOYSA-N
Synonym Methyloxalacetic Acid Diethyl Ester, 2-Ethoxalylpropionic Acid Ethyl Ester, Ethyl 2-Ethoxalylpropionate
PubChem CID 97750
ChEBI CHEBI:16879
IUPAC Name diethyl 2-methyl-3-oxobutanedioate
SMILES CCOC(=O)C(C)C(=O)C(=O)OCC

Specifications

Color Yellow
Boiling Point 133°C
Quantity 25 g
Formula Weight 202.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Methyloxalacetate