missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Methylmalonate 98.0+%, TCI America™

Catalog No. M036625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M036625ML 25 mL
M0366100ML 100 mL
M0366500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M036625ML Supplier TCI America Supplier No. M036625ML

Chemical Identifiers

CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009162
InChI Key UPQZOUHVTJNGFK-UHFFFAOYSA-N
Synonym diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl
PubChem CID 11857
IUPAC Name diethyl 2-methylpropanedioate
SMILES CCOC(=O)C(C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 199°C
Quantity 25 mL
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Methylmalonate