missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylmalonate, 99%

Catalog No. AC155141000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155141000 100 g Glass bottle
AC155145000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155141000 Supplier Thermo Scientific Chemicals Supplier No. 155141000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 609-08-5 | C8H14O4 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00009162
InChI Key UPQZOUHVTJNGFK-UHFFFAOYSA-N
Synonym diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl
PubChem CID 11857
IUPAC Name diethyl 2-methylpropanedioate
SMILES CCOC(=O)C(C)C(=O)OCC

Specifications

Color Colorless
Density 1.0100g/mL
Boiling Point 198°C to 199°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.414
Linear Formula C2H5O2CCH(CH3)CO2C2H5
Quantity 100 g
Beilstein 02, 629
Fieser 05,216
Specific Gravity 1.01
Solubility Information Solubility in water: immiscible
Formula Weight 174.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl methylmalonate, 99%
Show More Show Less
EINECSNumber 210-175-7
TSCA TSCA

RUO – Research Use Only