missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylmalonate, 99%

Catalog No. AAA1207818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1207818 50 g
AAA1207830 250 g
AAA120780B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1207818 Supplier Thermo Scientific Chemicals Supplier No. A1207818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 609-08-5 | C8H14O4 | 174.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009162
InChI Key UPQZOUHVTJNGFK-UHFFFAOYSA-N
Synonym diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl
PubChem CID 11857
IUPAC Name diethyl 2-methylpropanedioate
SMILES CCOC(=O)C(C)C(=O)OCC

Specifications

Density 1.02
Boiling Point 198°C to 199°C
Flash Point 85°C (185°F)
Refractive Index 1.413
Quantity 50 g
Beilstein 637028
Formula Weight 174.2
Percent Purity 99%
Chemical Name or Material Diethyl methylmalonate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-175-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only