missing translation for 'onlineSavingsMsg'
Learn More

Diethyl meso-2,5-dibromoadipate, 98%

Catalog No. AAH2634706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2634706 5 g
AAH2634714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2634706 Supplier Thermo Scientific Chemicals Supplier No. H2634706
Only null left

CAS: 869-10-3 | C10H16Br2O4 | 360.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 869-10-3
Molecular Formula C10H16Br2O4
Molecular Weight (g/mol) 360.04
MDL Number MFCD00075379
InChI Key UBCNJHBDCUBIPB-UHFFFAOYNA-N
Synonym diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate
PubChem CID 241588
IUPAC Name diethyl 2,5-dibromohexanedioate
SMILES CCOC(=O)C(Br)CCC(Br)C(=O)OCC

Specifications

Melting Point 64°C to 67°C
Quantity 5 g
UN Number UN3261
Sensitivity Light sensitive
Formula Weight 360.04
Percent Purity 98%
Chemical Name or Material Diethyl meso-2,5-dibromoadipate
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only