missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Malonate, TCI America™

Catalog No. p-7131938
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M002925ML 25 mL
M0029500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M002925ML Supplier TCI America Supplier No. M002925ML
Only null left

Chemical Identifiers

CAS 105-53-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00009195
InChI Key IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Synonym diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
PubChem CID 7761
IUPAC Name diethyl propanedioate
SMILES CCOC(=O)CC(=O)OCC

Specifications

Melting Point -50°C
Color Colorless
Boiling Point 199°C
Quantity 25 mL
Formula Weight 160.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Malonate
EINECSNumber (2)-0913
RTECSNumber OO0700000
TSCA Yes