missing translation for 'onlineSavingsMsg'
Learn More

Diethyl malonate, 99%

CAS: 105-53-3 | C7H12O4 | 160.169 g/mol

Supplier:  Thermo Scientific Chemicals A154680C

 View more versions of this product

Catalog No. AAA154680C


Only null left
Explore available promotions

Description

Description

Diethyl malonate is used in organic synthesis for the preparation of alpha-aryl malonates, mono-substituted and di-substituted acetic acid, barbiturates and artificial flavorings. It is also involved in the synthesis of pharmaceuticals like chloroquine, butazolidin and barbital. It acts as intermediate in the synthesis of vitamin B1, vitamin B6, non-steroidal anti-inflammatory agents agrochemicals and perfumes. In Knoevenagel condensation reaction, it reacts with benzaldehyde to get diethyl benzylidenemalonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl malonate is used in organic synthesis for the preparation of alpha-aryl malonates, mono-substituted and di-substituted acetic acid, barbiturates and artificial flavorings. It is also involved in the synthesis of pharmaceuticals like chloroquine, butazolidin and barbital. It acts as intermediate in the synthesis of vitamin B1, vitamin B6, non-steroidal anti-inflammatory agents agrochemicals and perfumes. In Knoevenagel condensation reaction, it reacts with benzaldehyde to get diethyl benzylidenemalonate.

Solubility
Miscible with ethyl alcohol, ether, chloroform and benzene. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications

Specifications

Diethyl malonate
-50°C
198°C to 200°C
99%
C7H12O4
MFCD00009195
774687
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
diethyl propanedioate
7761
99%
105-53-3
1.055
90°C (194°F)
Pleasant
1.414
10,000 g
14,3823
Miscible with ethyl alcohol,ether,chloroform and benzene. Slightly miscible with water.
CCOC(=O)CC(=O)OCC
160.169
160.17
Liquid

Chemical Identifiers

105-53-3
160.169
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
7761
CCOC(=O)CC(=O)OCC
C7H12O4
MFCD00009195
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
diethyl propanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 203-305-9

RTECSNumber : OO0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only