missing translation for 'onlineSavingsMsg'
Learn More

Diethyl malonate, 99+%

Catalog No. p-4258952
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114450250 25 g Glass bottle
AC114455000 500 g Glass bottle
AC114450010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114450250 Supplier Thermo Scientific Chemicals Supplier No. 114450250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-53-3 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-53-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
InChI Key IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Synonym diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
PubChem CID 7761
IUPAC Name diethyl propanedioate
SMILES CCOC(=O)CC(=O)OCC

Specifications

Melting Point -50.0°C
Density 1.0550g/mL
Boiling Point 199.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4130 to 1.4150
Linear Formula C2H5O2CCH2CO2C2H5
Quantity 25 g
Beilstein 02,573
Fieser 10,182; 13,69
Merck Index 15,3878
Specific Gravity 1.055
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone
Viscosity 2.14 mPa.s (10°C)
Formula Weight 160.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diethyl malonate
Show More Show Less
EINECSNumber 203-305-9

RUO – Research Use Only