missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Maleate 90.0+%, TCI America™

Catalog No. M0010500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0010500ML 500 mL
M001025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0010500ML Supplier TCI America Supplier No. M0010500ML

Chemical Identifiers

CAS 141-05-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009191
InChI Key IEPRKVQEAMIZSS-WAYWQWQTSA-N
Synonym diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
PubChem CID 5271566
ChEBI CHEBI:68508
IUPAC Name diethyl (Z)-but-2-enedioate
SMILES CCOC(=O)C=CC(=O)OCC

Specifications

Melting Point -10°C
Color Colorless
Boiling Point 225°C
Quantity 500 mL
Formula Weight 172.18
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Maleate
EINECSNumber (2)-1107&(2)-1142
RTECSNumber ON1225000
TSCA Yes