missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl maleate, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 141-05-9 |
| Density | 1.0600g/mL |
| Boiling Point | 225.0°C |
| Flash Point | 93°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C8H12O4 |
| Linear Formula | C2H5O2CCH=CHCO2C2H5 |
| Refractive Index | 1.4380 to 1.4420 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement: Wear eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 205-451-9 |
| RTECSNumber | ON1225000 |
| TSCA | TSCA |
RUO – Research Use Only