missing translation for 'onlineSavingsMsg'
Learn More

Diethyl maleate, 97%

CAS: 141-05-9 | C8H12O4 | 172.18 g/mol

Supplier:  Thermo Scientific Chemicals L038760B

 View more versions of this product

Catalog No. AAL038760B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl maleate
-11°C
221°C to 224°C
Pleasant
1.44
1000 g
14,3123
IEPRKVQEAMIZSS-WAYWQWQTSA-N
diethyl (Z)-but-2-enedioate
5271566
172.18
141-05-9
1.064
93°C (199°F)
C8H12O4
MFCD00009191
1100825
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
CCOC(=O)C=CC(=O)OCC
172.18
CHEBI:68508
97%

Chemical Identifiers

141-05-9
172.18
IEPRKVQEAMIZSS-WAYWQWQTSA-N
5271566
diethyl (Z)-but-2-enedioate
C8H12O4
MFCD00009191
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
CHEBI:68508
CCOC(=O)C=CC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317-H227
May cause an allergic skin reaction.
Combustible liquid.

P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c

H227-H317

EINECSNumber : 205-451-9

RTECSNumber : ON1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only