missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Diethyl L-tartrate, 99+%
CAS: 87-91-2 | C8H14O6 | 206.19 g/mol
$244.88 - $244.88
Chemical Identifiers
| CAS | 87-91-2 |
|---|---|
| Molecular Formula | C8H14O6 |
| Molecular Weight (g/mol) | 206.19 |
| MDL Number | MFCD00009143 |
| InChI Key | YSAVZVORKRDODB-UHFFFAOYNA-N |
| Synonym | diethyl l-+-tartrate, 2r,3r-diethyl 2,3-dihydroxysuccinate, l +-diethyl l-tartrate, +-diethyl l-tartrate, diethyl l-tartrate, diethyl-l-tartrate, unii-oq72cpy58z, l-+-tartaric acid diethyl ester, diethyl l-tartarate, tartaric acid, diethyl ester, r,r |
| PubChem CID | 6993580 |
| IUPAC Name | diethyl (2R,3R)-2,3-dihydroxybutanedioate |
| SMILES | CCOC(=O)C(O)C(O)C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC168965000
|
Thermo Scientific Chemicals
168965000 |
500 mL | Glass bottle |
Each for $244.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 87-91-2 | |
| 206.19 | |
| YSAVZVORKRDODB-UHFFFAOYNA-N | |
| 6993580 | |
| CCOC(=O)C(O)C(O)C(=O)OCC |
| C8H14O6 | |
| MFCD00009143 | |
| diethyl l-+-tartrate, 2r,3r-diethyl 2,3-dihydroxysuccinate, l +-diethyl l-tartrate, +-diethyl l-tartrate, diethyl l-tartrate, diethyl-l-tartrate, unii-oq72cpy58z, l-+-tartaric acid diethyl ester, diethyl l-tartarate, tartaric acid, diethyl ester, r,r | |
| diethyl (2R,3R)-2,3-dihydroxybutanedioate |
Specifications
| 87-91-2 | |
| 1.2000g/mL | |
| 93°C | |
| 99% min. (GC) | |
| C8H14O6 | |
| [-CH(OH)CO2C2H5]2 | |
| 500 mL | |
| 11,181; 13,276; 16,54 | |
| 15, 3150 | |
| + 7.5 | |
| Solubility in water: insoluble. Other solubilities: soluble in organic solvents | |
| CCOC(=O)C(O)C(O)C(=O)OCC | |
| 206.19 | |
| 206.19 | |
| Viscous Liquid |
| 17°C | |
| 280°C | |
| Authentic | |
| Glass bottle | |
| 1.4450 to 1.447 | |
| MFCD00009143 | |
| 03, 512 | |
| 1.2 | |
| +7.50° (20°C neat) | |
| diethyl l-+-tartrate, 2r,3r-diethyl 2,3-dihydroxysuccinate, l +-diethyl l-tartrate, +-diethyl l-tartrate, diethyl l-tartrate, diethyl-l-tartrate, unii-oq72cpy58z, l-+-tartaric acid diethyl ester, diethyl l-tartarate, tartaric acid, diethyl ester, r,r | |
| YSAVZVORKRDODB-UHFFFAOYNA-N | |
| diethyl (2R,3R)-2,3-dihydroxybutanedioate | |
| 6993580 | |
| 99+% | |
| (+)-Diethyl L-tartrate, 99+% |
Safety and Handling
EINECSNumber : 201-783-3
TSCA : TSCA
RUO – Research Use Only