missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diethyl L-tartrate, 99+%

Catalog No. AC168965000
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168965000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC168965000 Supplier Thermo Scientific Chemicals Supplier No. 168965000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-91-2 | C8H14O6 | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-91-2
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00009143
InChI Key YSAVZVORKRDODB-UHFFFAOYNA-N
Synonym diethyl l-+-tartrate, 2r,3r-diethyl 2,3-dihydroxysuccinate, l +-diethyl l-tartrate, +-diethyl l-tartrate, diethyl l-tartrate, diethyl-l-tartrate, unii-oq72cpy58z, l-+-tartaric acid diethyl ester, diethyl l-tartarate, tartaric acid, diethyl ester, r,r
PubChem CID 6993580
IUPAC Name diethyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)C(O)C(O)C(=O)OCC

Specifications

Melting Point 17°C
Density 1.2000g/mL
Boiling Point 280°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2C2H5]2
Refractive Index 1.4450 to 1.447
Quantity 500 mL
Beilstein 03, 512
Fieser 11,181; 13,276; 16,54
Specific Gravity 1.2
Merck Index 15, 3150
Specific Rotation Condition +7.50° (20°C neat)
Specific Rotation + 7.5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 206.19
Percent Purity 99+%
Physical Form Viscous Liquid
Chemical Name or Material (+)-Diethyl L-tartrate, 99+%
Show More Show Less
EINECSNumber 201-783-3
TSCA TSCA

RUO – Research Use Only