missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl(isopropyl)silanol 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 146.31 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl(isopropyl)silanol |
Safety and Handling
| TSCA | No |