missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Isobutylmalonate 98.0+%, TCI America™

Catalog No. i0251500ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0251500ML 500 mL
I025125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0251500ML Supplier TCI America Supplier No. I0251500ML
Only null left

Chemical Identifiers

CAS 10203-58-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00026869
InChI Key OFRFGNSZCYDFOH-UHFFFAOYSA-N
Synonym diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate
PubChem CID 25047
IUPAC Name diethyl 2-(2-methylpropyl)propanedioate
SMILES CCOC(=O)C(CC(C)C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 225°C
Quantity 500 mL
Formula Weight 216.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Isobutylmalonate
EINECSNumber (2)-0879
TSCA Yes