missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isobutylmalonate, 98%

Catalog No. aal0920206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0920206 5 g
AAL0920214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0920206 Supplier Thermo Scientific Chemicals Supplier No. L0920206
Only null left

CAS: 10203-58-4 | C11H20O4 | 216.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10203-58-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00026869
InChI Key OFRFGNSZCYDFOH-UHFFFAOYSA-N
Synonym diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate
PubChem CID 25047
IUPAC Name diethyl 2-(2-methylpropyl)propanedioate
SMILES CCOC(=O)C(CC(C)C)C(=O)OCC

Specifications

Density 0.97
Boiling Point 225°C
Flash Point >100°C (212°F)
Refractive Index 1.422
Quantity 5 g
Beilstein 1103030
Formula Weight 216.28
Percent Purity 98%
Chemical Name or Material Diethyl isobutylmalonate
EINECSNumber 233-504-6
RTECSNumber TY1850200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only