missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodiacetate, 98%

Catalog No. AC407321000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407321000 100 g Glass bottle
AC407320250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407321000 Supplier Thermo Scientific Chemicals Supplier No. 407321000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6290-05-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00041925
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
PubChem CID 80502
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
SMILES CCOC(=O)CNCC(=O)OCC

Specifications

Color Yellow
Density 1.0800g/mL
Boiling Point 248.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370
Linear Formula HN(CH2COOC2H5)2
Quantity 100 g
Specific Gravity 1.08
Formula Weight 189.21
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Diethyl iminodiacetate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-533-6
TSCA TSCA

RUO – Research Use Only