missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Hydroxymethyl)phosphonate 97.0+%, TCI America™

Catalog No. D381325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D381325G 25 g
D3813100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D381325G Supplier TCI America Supplier No. D381325G

Chemical Identifiers

CAS 3084-40-0
Molecular Formula C5H13O4P
Molecular Weight (g/mol) 168.129
MDL Number MFCD00014418
InChI Key RWIGWWBLTJLKMK-UHFFFAOYSA-N
Synonym diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
PubChem CID 76513
IUPAC Name diethoxyphosphorylmethanol
SMILES CCOP(=O)(CO)OCC

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
Formula Weight 168.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Hydroxymethyl)phosphonate
TSCA Yes
Recommended Storage Refrigerator