missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (hydroxymethyl)phosphonate, 97%

Catalog No. AC320950250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC320950250 25 g Glass bottle
AC320950050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC320950250 Supplier Thermo Scientific Chemicals Supplier No. 320950250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3084-40-0 | C5H13O4P | 168.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 3084-40-0
Molecular Formula C5H13O4P
Molecular Weight (g/mol) 168.13
MDL Number MFCD00014418
InChI Key RWIGWWBLTJLKMK-UHFFFAOYSA-N
Synonym diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
PubChem CID 76513
IUPAC Name diethoxyphosphorylmethanol
SMILES CCOP(=O)(CO)OCC

Specifications

Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 124.0°C to 126.0°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4370 to 1.4410
Linear Formula HOCH2P(O)(OC2H5)2
Quantity 25 g
Specific Gravity 1.14
Formula Weight 168.13
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material Diethyl (hydroxymethyl)phosphonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-391-6

RUO – Research Use Only