missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Glutarate 98.0+%, TCI America™
$65.24 - $425.00
Chemical Identifiers
| CAS | 818-38-2 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.22 |
| MDL Number | MFCD00009212 |
| InChI Key | OUWSNHWQZPEFEX-UHFFFAOYSA-N |
| Synonym | diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters |
| PubChem CID | 13163 |
| ChEBI | CHEBI:87319 |
| IUPAC Name | 1,5-diethyl pentanedioate |
| SMILES | CCOC(=O)CCCC(=O)OCC |
Chemical Identifiers
| 818-38-2 | |
| 188.22 | |
| OUWSNHWQZPEFEX-UHFFFAOYSA-N | |
| 13163 | |
| 1,5-diethyl pentanedioate |
| C9H16O4 | |
| MFCD00009212 | |
| diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters | |
| CHEBI:87319 | |
| CCOC(=O)CCCC(=O)OCC |
Specifications
| 818-38-2 | |
| 230°C | |
| MFCD00009212 | |
| diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters | |
| CCOC(=O)CCCC(=O)OCC | |
| 188.22 | |
| CHEBI:87319 | |
| ≥98.0% (GC) | |
| Diethyl Glutarate |
| Yellow | |
| C9H16O4 | |
| 25 g | |
| OUWSNHWQZPEFEX-UHFFFAOYSA-N | |
| 1,5-diethyl pentanedioate | |
| 13163 | |
| 188.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3088
TSCA : Yes