missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Fumarate 98.0+%, TCI America™

Catalog No. F006825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F006825ML 25 mL
F0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F006825ML Supplier TCI America Supplier No. F006825ML

Chemical Identifiers

CAS 623-91-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00064455
InChI Key IEPRKVQEAMIZSS-AATRIKPKSA-N
Synonym diethyl fumarate, fumaric acid, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, fumaric acid diethyl ester, diethyl e-but-2-enedioate, 2-butenedioic acid e-, diethyl ester, trans-2-butenedioic acid diethyl ester, 2-butenedioic acid 2e-, diethyl ester, unii-5wbu5a3e8a
PubChem CID 638144
ChEBI CHEBI:87388
IUPAC Name 1,4-diethyl (2E)-but-2-enedioate
SMILES CCOC(=O)\C=C\C(=O)OCC

Specifications

Melting Point 2°C
Color Colorless
Boiling Point 219°C
Quantity 25 mL
Formula Weight 172.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Fumarate
EINECSNumber (2)-1099&(2)-1142
RTECSNumber EM5950000
TSCA Yes