missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Fluoromalonate 95.0+%, TCI America™

Catalog No. p-7129589
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02831G 1 g
F02835G 5 g
F028325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F02831G Supplier TCI America Supplier No. F02831G
Only null left

Chemical Identifiers

CAS 685-88-1
Molecular Formula C7H11FO4
Molecular Weight (g/mol) 178.159
MDL Number MFCD00009139
InChI Key GOWQBFVDZPZZFA-UHFFFAOYSA-N
PubChem CID 12702
IUPAC Name diethyl 2-fluoropropanedioate
SMILES CCOC(=O)C(C(=O)OCC)F

Specifications

Color Colorless
Boiling Point 122°C
Quantity 1 g
UN Number 1760
Synonym diethyl fluoromalonate, diethyl 2-fluoromalonate, fluoromalonic acid diethyl ester, 1,3-diethyl 2-fluoropropanedioate, diethylfluoromalonate, 2-fluoro-malonic acid diethyl ester, diethyl2-fluoromalonate, diethyl fluoropropanedioate, propanedioic acid, fluoro-, diethyl ester, malonic acid, fluoro-, diethyl ester
Formula Weight 178.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Fluoromalonate
RTECSNumber OO1670000
TSCA No