missing translation for 'onlineSavingsMsg'
Learn More

Diethyl fluoromalonate, 97%

Catalog No. AAB2076603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2076603 1 g
AAB2076606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2076603 Supplier Thermo Scientific Chemicals Supplier No. B2076603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 685-88-1 | C7H11FO4 | 178.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 685-88-1
Molecular Formula C7H11FO4
Molecular Weight (g/mol) 178.159
MDL Number MFCD00009139
InChI Key GOWQBFVDZPZZFA-UHFFFAOYSA-N
Synonym diethyl fluoromalonate, diethyl 2-fluoromalonate, fluoromalonic acid diethyl ester, 1,3-diethyl 2-fluoropropanedioate, diethylfluoromalonate, 2-fluoro-malonic acid diethyl ester, diethyl2-fluoromalonate, diethyl fluoropropanedioate, propanedioic acid, fluoro-, diethyl ester, malonic acid, fluoro-, diethyl ester
PubChem CID 12702
IUPAC Name diethyl 2-fluoropropanedioate
SMILES CCOC(=O)C(C(=O)OCC)F

Specifications

Density 1.129
Boiling Point 121°C to 122°C (30 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.407
Quantity 1 g
UN Number UN1760
Beilstein 1775686
Formula Weight 178.16
Percent Purity 97%
Chemical Name or Material Diethyl fluoromalonate
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 211-684-7
RTECSNumber OO1670000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only