missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ethylmalonate 98.0+%, TCI America™
$39.14 - $143.54
Chemical Identifiers
| CAS | 133-13-1 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.223 |
| MDL Number | MFCD00009167 |
| InChI Key | VQAZCUCWHIIFGE-UHFFFAOYSA-N |
| Synonym | diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate |
| PubChem CID | 8610 |
| IUPAC Name | diethyl 2-ethylpropanedioate |
| SMILES | CCC(C(=O)OCC)C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E013825ML
|
TCI America
E013825ML |
25 mL |
Each for $39.14
|
|
|||||
|
E0138500ML
|
TCI America
E0138500ML |
500 mL |
Each for $143.54
|
|
|||||
Chemical Identifiers
| 133-13-1 | |
| 188.223 | |
| VQAZCUCWHIIFGE-UHFFFAOYSA-N | |
| 8610 | |
| CCC(C(=O)OCC)C(=O)OCC |
| C9H16O4 | |
| MFCD00009167 | |
| diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate | |
| diethyl 2-ethylpropanedioate |
Specifications
| 133-13-1 | |
| 211°C | |
| MFCD00009167 | |
| diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate | |
| CCC(C(=O)OCC)C(=O)OCC | |
| 188.223 | |
| 188.22 | |
| Liquid |
| Colorless | |
| C9H16O4 | |
| 25 mL | |
| VQAZCUCWHIIFGE-UHFFFAOYSA-N | |
| diethyl 2-ethylpropanedioate | |
| 8610 | |
| ≥98.0% (GC) | |
| Diethyl Ethylmalonate |
Safety and Handling
EINECSNumber : (9)-0264
TSCA : Yes