missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ethylmalonate 98.0+%, TCI America™

Catalog No. E0138500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0138500ML 500 mL
E013825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0138500ML Supplier TCI America Supplier No. E0138500ML

Chemical Identifiers

CAS 133-13-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00009167
InChI Key VQAZCUCWHIIFGE-UHFFFAOYSA-N
Synonym diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate
PubChem CID 8610
IUPAC Name diethyl 2-ethylpropanedioate
SMILES CCC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 211°C
Quantity 500 mL
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Ethylmalonate
EINECSNumber (9)-0264
TSCA Yes