missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ethoxymethylenemalonate 98.0+%, TCI America™

Catalog No. E0255500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0255500ML 500 mL
E025525ML 25 mL
E0255100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0255500ML Supplier TCI America Supplier No. E0255500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-13-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009148
InChI Key LTMHNWPUDSTBKD-UHFFFAOYSA-N
Synonym diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate
PubChem CID 6871
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
SMILES CCOC=C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 166°C
Quantity 500 mL
Formula Weight 216.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Ethoxymethylenemalonate
EINECSNumber (9)-0561
RTECSNumber OO1100000
TSCA Yes