missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ethoxymethylenemalonate, 99+%

Catalog No. p-108961
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC114341000 Supplier Thermo Scientific Chemicals Supplier No. 114341000

CAS: 87-13-8 | C10H16O5 | 216.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-13-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00009148
InChI Key LTMHNWPUDSTBKD-UHFFFAOYSA-N
Synonym diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate
PubChem CID 6871
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
SMILES CCOC=C(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 279°C to 281°C
Flash Point 144°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.463
Linear Formula C2H5OCH=C(CO2C2H5)2
Quantity 100 g
Beilstein 03, 469
Fieser 05,216
Specific Gravity 1.08
Solubility Information Solubility in water: insoluble
Formula Weight 216.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diethyl ethoxymethylenemalonate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-725-7
RTECSNumber OO1100000
TSCA TSCA

RUO – Research Use Only