missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.8% (GC), Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -116°C |
| Appearance | Liquid |
| Density | 0.706g/mL at 25°C |
| Quantity | 5 L |
| Explosion Limit | 0.365 |
| Flash Point | −40°C (−40°F) |
| Ignition Point | 320°F |
| Packaging | Aluminum Bottle |
| Refractive Index | n20/D 1.3530 |
| Residue after Evaporation | 0 to 0.001 % |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261 |
| Precautionary Statement | H224-H302-H336 |
| DOTInformation | Class 3 - PG 1 - Diethyl ether |
| EINECSNumber | 200-467-2 |
| RTECSNumber | KI5775000 |