missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.8% (GC), Honeywell™ Riedel-de Haën™

Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

Supplier:  Honeywell-Riedel-de Haen 322034X5L

 View more versions of this product

Catalog No. 60046074


Only null left
Explore available promotions

Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Specifications

Diethyl Ether
Reag. Ph. Eur. reag. ISO
Liquid
34.6°C
0.365
320°F
n20/D 1.3530
C4H10O
MFCD00011646
1696894
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12g/mol
ACS/puriss. p.a.
-116°C
60-29-7
0.706g/mL at 25°C
4 x 5 L
−40°C (−40°F)
Aluminum Bottle
0 to 0.001 %
(CH3CH2)2O
UN1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
≥99.8% (GC)

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261

H224-H302-H336

DOTInformation : Class 3 - PG 1 - Diethyl ether

EINECSNumber : 200-467-2

RTECSNumber : KI5775000

SDS
Documents

Documents

Promotions

Promotions