missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.8% (GC), Honeywell™ Riedel-de Haën™
Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%
Supplier: Honeywell-Riedel-de Haen 3220325L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Diethyl Ether | |
| Reag. Ph. Eur. reag. ISO | |
| Liquid | |
| 34.6°C | |
| 0.365 | |
| 320°F | |
| n20/D 1.3530 | |
| C4H10O | |
| MFCD00011646 | |
| 1696894 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12g/mol | |
| ACS/puriss. p.a. |
| -116°C | |
| 60-29-7 | |
| 0.706g/mL at 25°C | |
| 2.5 L | |
| −40°C (−40°F) | |
| Glass Bottle | |
| 0 to 0.001 % | |
| (CH3CH2)2O | |
| UN1155 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| ≥99.8% (GC) |
Chemical Identifiers
| 60-29-7 | |
| 74.12 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| 3283 | |
| ethoxyethane |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Safety and Handling
P210-P261
H224-H302-H336
DOTInformation : Class 3 - PG 1 - Diethyl ether
EINECSNumber : 200-467-2
RTECSNumber : KI5775000