missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -116°C |
| Density | 0.706g/mL at 25°C |
| Quantity | 5 L |
| Explosion Limit | 0.365 |
| Flash Point | −45°C (−49°F) |
| Ignition Point | 320°F |
| Packaging | Aluminum Bottle |
| Physical Form | Liquid |
| Linear Formula | (CH3CH2)2O |
| UN Number | UN1155 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261 |
| Precautionary Statement | H224-H302-H336 |
| DOTInformation | class 3 - PG 1 - Diethyl ether |
| EINECSNumber | 200-467-2 |
| RTECSNumber | KI5775000 |