missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™

$1,250.58 - $1,250.58

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
E199-4
SDS Accessories View Documents
Fisher Chemical
FLE1994
4 L Amber Glass N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

-116°C
0.01 AU max. at 300nm, 0.01 AU max. at 350nm, 0.02 AU max. at 270nm, 0.08 AU max. at 250nm, 1.00 AU max. at 218nm
0.714g/cm³
4 L
Liquid
≥99.5 %
C2H5OC2H5
587mbar at 20°C
0.08% max.
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Pesticide
60-29-7
Colorless
34.6°C
Amber Glass
3ppm max.
C4H10O
MFCD00011646
0.2448 cP at 20°C
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.12
CHEBI:35702
≥99.5%
Ethyl Ether
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Wear personal protective equipment. Handle under an inert atmosphere. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid contact with skin, eyes and clothing. Do not breathe vapors or spray mist.

NFPA

Health:1
Flammability:4
Instability:1

SDS
missing translation for 'documents'

missing translation for 'documents'