missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, For pesticide residue analysis, Honeywell™ Riedel-de Haën™

Catalog No. p-7072046
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000524 6 x 1 L
6000525 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000524 Supplier Honeywell Chemicals Supplier No. 316716X1L
Only null left

CHROMASOLV™ for pesticide residue analysis,≥99.8%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Color Colorless
Density 0.706g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Linear Formula (CH3CH2)2O
Vapor Pressure 2311 hPa (60°C)
UN Number 1155
Beilstein 1696894
Formula Weight 74.12g/mol
Grade CHROMASOLV
Chemical Name or Material Diethyl Ether
Show More Show Less
Hazard Statement P210-P261
Precautionary Statement H224-H302-H336
EINECSNumber 200-467-2
RTECSNumber KI5775000