missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether For gas chromatography ECD and FID SupraSolv™ MilliporeSigma™

Catalog No. p-7020428
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1009312500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1009312500 Supplier Technidata Supplier No. 1.00931.2500
Only null left

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.3°C
Color Colorless
Density 0.71g/cm3 at 20°C
Quantity 2.5 L
Flash Point −40°C (−40°F)
Ignition Point 180°C (356°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 587 hPa (20°C)
UN Number 1155
Solubility Information Soluble in water: 69g/L at 20°C
Boiling Point 34.6°C
Formula Weight 74.12
Chemical Name or Material Diethyl Ether
Show More Show Less
Hazard Category H224: Extremely flammable liquid and vapour.
Hazard Statement H302: Harmful if swallowed.
Precautionary Statement H336: May cause drowsiness or dizziness.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.