missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023036
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023036 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023036 Supplier Merck Emd Millipore Supplier No. RTC00007720ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 60-29-7
Melting Point -116°C (literature)
Density 0.706 g/mL (at 25°C (literature))
Boiling Point 34.6°C (literature)
Flash Point -40°C (Not Applicable)
Molecular Formula C4H10O
Refractive Index n20/D 1.353 (literature)
Linear Formula (CH3CH2)2 O
Quantity 20 mL
MDL Number MFCD00011646
Synonym Diethyl ether; Ether; Ethyl ether
Vapor Density 2.6 (vs air)
Formula Weight 74.12
Grade Certified Reference Material
Show More Show Less