missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT
Description
Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 116°C |
| Density | 0.713 |
| Quantity | 50 g |
| Flash Point | -45°C (-49°F) |
| Physical Form | Liquid |
| Refractive Index | 1.3525 |
| Assay Percent Range | 99% min. |
| Linear Formula | (C2H5)2O |
| UN Number | UN1155 |
| Merck Index | 14,3806 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H224-H302-H305-H335-H336-H373 |
| missing translation for 'healthHazard2' | GHS H Statement H224-H302-H336 |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER |
| EINECSNumber | 200-467-2 |
| RTECSNumber | KI5775000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only