missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%, Honeywell Riedel-de Haën™

Catalog No. 60046383
Change view
Click to view available options
Quantity:
250 mL
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046383 6 x 1 L Glass Bottle
60046382 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046383 Supplier Honeywell Inc Supplier No. 3461366X1L
Only null left

ACS Reagent, contains BHT as inhibitor, anhydrous,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form anhydrous
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99%
Grade ACS
Assay (GC)
Chemical Name or Material Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
EINECSNumber 200-467-2
Recommended Storage Room Temp