missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%, Honeywell Riedel-de Haën™
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -116°C (literature) |
| CAS Max % | ≤100.0000% |
| Density | 0.706 g/mL at 25 °C(lit.) |
| Quantity | 4 x 4 L |
| Flash Point | −45°C |
| Packaging | Glass Bottle |
| Physical Form | anhydrous |
| Linear Formula | (CH3CH2)2O |
| UN Number | UN1155 |
| Beilstein | 1696894 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether |
| EINECSNumber | 200-467-2 |
| Recommended Storage | Room Temp |