Learn More
Diethyl ether, 99+%, ACS reagent, anhydrous, BHT stab., meets req. of Reag.Ph.Eur.
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Supplier: Thermo Scientific Chemicals 410065000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Diethyl Ether | |
| Anhydrous, stabilized with BHT | |
| 0.7140g/mL | |
| 500 g | |
| Glass Bottle | |
| 99% min. (GC) | |
| 0.0002 meq/g max | |
| (CH3CH2)2O | |
| 0.2448 mPa.s (20°C) | |
| 11,3762 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12 | |
| ACS Reagent |
| -116.0°C | |
| 60-29-7 | |
| 34.6°C | |
| −45°C | |
| 1.3510 to 1.3530 | |
| 0.714 | |
| C4H10O | |
| MFCD00011646 | |
| 01,314 | |
| 15,3861 | |
| Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| 99+% |
Chemical Identifiers
| 60-29-7 | |
| 74.12 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| 3283 | |
| ethoxyethane |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No
GHS Signal Word: Danger
EINECSNumber : 200-467-2
RUO – Research Use Only