missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Dodecanedioate 97.0+%, TCI America™

Catalog No. d248625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D248625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D248625G Supplier TCI America Supplier No. D248625G
Only null left

Chemical Identifiers

CAS 10471-28-0
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.41
MDL Number MFCD00009204
InChI Key AFYNPRWLOKYLDU-UHFFFAOYSA-N
Synonym Dodecanedioic Acid Diethyl Ester
PubChem CID 82650
IUPAC Name 1,12-diethyl dodecanedioate
SMILES CCOC(=O)CCCCCCCCCCC(=O)OCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 286.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Dodecanedioate
EINECSNumber (2)-0879
TSCA No