missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Dipropylmalonate 97.0+%, TCI America™

Catalog No. p-7127483
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D276225ML 25 mL
D2762500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D276225ML Supplier TCI America Supplier No. D276225ML

Chemical Identifiers

CAS 6065-63-0
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.331
MDL Number MFCD00026840
InChI Key NNDOHYGFLASMFR-UHFFFAOYSA-N
PubChem CID 80168
IUPAC Name diethyl 2,2-dipropylpropanedioate
SMILES CCCC(CCC)(C(=O)OCC)C(=O)OCC

Specifications

Color Yellow
Boiling Point 249°C
Quantity 25 mL
Synonym diethyl dipropylmalonate, diethyl 2,2-dipropylmalonate, diethyl-dipropylmalonate, diethyldipropylmalonate, diethyl di-n-propylmalonate, 1,3-diethyl 2,2-dipropylpropanedioate, propanedioic acid, dipropyl-, diethyl ester, malonic acid, dipropyl-, diethyl ester, acmc-209mla, diethyl2,2-dipropylmalonate
Formula Weight 244.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Dipropylmalonate
EINECSNumber (2)-0879
TSCA No