missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Diethylmalonate 97.0+%, TCI America™

Catalog No. D086725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D086725ML 25 mL
D0867500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D086725ML Supplier TCI America Supplier No. D086725ML

Chemical Identifiers

CAS 77-25-8
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00009132
InChI Key ZKBBUZRGPULIRN-UHFFFAOYSA-N
Synonym diethyl diethylmalonate, diethyl 2,2-diethylmalonate, diethylmalonic ester, propanedioic acid, diethyl-, diethyl ester, diethylmalonate diethyl ester, diethylmalonic acid diethyl ester, diethylmalonic acid, diethyl ester, unii-ceh13944yq, 1,3-diethyl 2,2-diethylpropanedioate, diethyl diethylmalonate diethyl
PubChem CID 66165
IUPAC Name diethyl 2,2-diethylpropanedioate
SMILES CCC(CC)(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 216.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Diethylmalonate
EINECSNumber (2)-0914&(2)-0879
TSCA Yes