missing translation for 'onlineSavingsMsg'
Learn More

Diethyl dibromomalonate, 96%

Catalog No. AAA1150822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150822 100 g
AAA1150806 5 g
AAA1150814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1150822 Supplier Thermo Scientific Chemicals Supplier No. A1150822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 631-22-1 | C7H10Br2O4 | 317.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 631-22-1
Molecular Formula C7H10Br2O4
Molecular Weight (g/mol) 317.96
MDL Number MFCD00015154
InChI Key PFZYFZRUPFUEOB-UHFFFAOYSA-N
PubChem CID 69427
IUPAC Name diethyl 2,2-dibromopropanedioate
SMILES CCOC(=O)C(Br)(Br)C(=O)OCC

Specifications

Density 1.68
Boiling Point 108°C to 110°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.485
Quantity 100 g
UN Number UN1760
Beilstein 1783879
Synonym diethyl dibromomalonate, diethyl 2,2-dibromomalonate, ethyl dibromomalonate, propanedioic acid, dibromo-, diethyl ester, malonic acid, dibromo-, diethyl ester, propanedioic acid, 2,2-dibromo-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibromopropanedioate, diethyldibromomalonate, acmc-1cuuu, diethyl2,2-dibromomalonate
Formula Weight 317.96
Percent Purity 97%
Chemical Name or Material Diethyl dibromomalonate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 211-151-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only