missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Diallylmalonate 98.0+%, TCI America™

Catalog No. D118325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D118325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D118325ML Supplier TCI America Supplier No. D118325ML
Only null left

Chemical Identifiers

CAS 3195-24-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD00009126
InChI Key LYUUVYQGUMRKOV-UHFFFAOYSA-N
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
PubChem CID 76664
IUPAC Name diethyl 2,2-bis(prop-2-enyl)propanedioate
SMILES CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
Formula Weight 240.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Diallylmalonate