missing translation for 'onlineSavingsMsg'
Learn More

Diethyl diallylmalonate, 97+%

Catalog No. p-7082160
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1246009 10 g
AAA1246018 50 g
AAA1246030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1246009 Supplier Thermo Scientific Chemicals Supplier No. A1246009
Only null left

CAS: 3195-24-2 | C13H20O4 | 240.299 g/mol

Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke?s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

Solubility
Sparingly Soluble in water. (0.72 g/L) (25°C)

Notes
Store under cool dry place. Ensure good ventilation.

Chemical Identifiers

CAS 3195-24-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD00009126
InChI Key LYUUVYQGUMRKOV-UHFFFAOYSA-N
PubChem CID 76664
IUPAC Name diethyl 2,2-bis(prop-2-enyl)propanedioate
SMILES CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Specifications

Density 0.993
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.446
Quantity 10 g
Beilstein 1790529
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
Solubility Information Sparingly Soluble in water. (0.72g/L) (25°C)
Formula Weight 240.3
Percent Purity 98%
Chemical Name or Material Diethyl diallylmalonate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-696-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only