missing translation for 'onlineSavingsMsg'
Learn More

Diethyl di-n-butylmalonate, 98+%, Thermo Scientific Chemicals

Catalog No. AAL0990214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0990214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0990214 Supplier Thermo Scientific Chemicals Supplier No. L0990214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 596-75-8 | Molecular Formula: C15H28O4 | Molecular Weight: 272.385 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 596-75-8
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.385
MDL Number MFCD00026842
InChI Key WHKKUUPZLWUOIW-UHFFFAOYSA-N
Synonym diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
PubChem CID 69000
IUPAC Name diethyl 2,2-dibutylpropanedioate
SMILES CCCCC(CCCC)(C(=O)OCC)C(=O)OCC

Specifications

Density 0.95
Boiling Point 150°C (12 mmHg)
Refractive Index 1.433
Quantity 25 g
Beilstein 518421
Formula Weight 272.39
Percent Purity ≥98%
Chemical Name or Material Diethyl di-n-butylmalonate
EINECSNumber 209-890-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only