missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Cyanomethylphosphonate, TCI America™

Catalog No. C14305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14305G 5 g
C143025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14305G Supplier TCI America Supplier No. C14305G

Chemical Identifiers

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00001893
InChI Key KWMBADTWRIGGGG-UHFFFAOYSA-N
Synonym diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
PubChem CID 75676
IUPAC Name diethyl (cyanomethyl)phosphonate
SMILES CCOP(=O)(CC#N)OCC

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 g
UN Number 3278
Formula Weight 177.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Cyanomethylphosphonate
EINECSNumber (2)-3973
TSCA No