missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanomethylphosphonate, 96%

Catalog No. AC114272500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114272500 250 g Glass bottle
AC114270100 10 g Glass bottle
AC114270500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114272500 Supplier Thermo Scientific Chemicals Supplier No. 114272500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2537-48-6 | C6H12NO3P | 177.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00001893
InChI Key KWMBADTWRIGGGG-UHFFFAOYSA-N
Synonym diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
PubChem CID 75676
IUPAC Name 2-diethoxyphosphorylacetonitrile
SMILES CCOP(=O)(CC#N)OCC

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 101.0°C to 102.0°C (0.4 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4312 to 1.4332
Linear Formula (C2H5O)2P(O)CH2CN
Quantity 250 g
Fieser 01,250; 02,130; 03,96; 04,150; 13,15
Specific Gravity 1.09
Formula Weight 177.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl cyanomethylphosphonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
EINECSNumber 219-806-

RUO – Research Use Only