missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chlorothiophosphate 97.0+%, TCI America™

Catalog No. D062425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D062425G 25 g
D0624500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D062425G Supplier TCI America Supplier No. D062425G
Only null left

Chemical Identifiers

CAS 2524-04-1
Molecular Formula C4H10ClO2PS
Molecular Weight (g/mol) 188.61
MDL Number MFCD00009088
InChI Key KMJJJTCKNZYTEY-UHFFFAOYSA-N
PubChem CID 17305
IUPAC Name chloro-diethoxy-sulfanylidene-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)Cl

Specifications

Boiling Point 51°C
Quantity 25 g
UN Number 2751
Synonym O,O′C-Diethyl Phosphorochloridothioate, O,O′C-Diethyl Phosphorothiochloridate, O,O′C-Diethylthiophosphoryl Chloride
Formula Weight 188.61
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethyl Chlorothiophosphate
EINECSNumber (2)-2939
RTECSNumber TD1780000
TSCA Yes